missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-4-phenylthiazole hydrobromide monohydrate, 99%, Thermo Scientific™ Product Code.: 11494107

2-Amino-4-phenylthiazole hydrobromide monohydrate, 99%, Thermo Scientific™

Product Code. 11494107
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11494107

Brand: Thermo Scientific Alfa Aesar A19542.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52253-69-7
Molecular Formula C9H11BrN2OS
Molecular Weight (g/mol) 275.164
MDL Number MFCD00150083
InChI Key SBVYKOGRUCDICV-UHFFFAOYSA-N
Synonym 2-amino-4-phenylthiazole hydrobromide monohydrate, 2-amino-4-phenylthiazole hbr h2o, 2-thiazolamine, 4-phenyl-, monohydrobromide, monohydrate, 4-phenylthiazol-2-amine hydrobromide hydrate, 2-amino-4-phenylthiazole hydrobromide hydrate, 4-phenyl-1,3-thiazol-2-amine hydrobromide hydrate, phenthiazamine hydrate hydrobromide, acmc-1aw4h, dsstox_cid_4503, dsstox_rid_77432
PubChem CID 40301
IUPAC Name 4-phenyl-1,3-thiazol-2-amine;hydrate;hydrobromide
SMILES C1=CC=C(C=C1)C2=CSC(=N2)N.O.Br

Specifications

Melting Point 180°C to 183°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 275.17 (257.15 Anhydrous)
Percent Purity 99%
Chemical Name or Material 2-Amino-4-phenylthiazole hydrobromide monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-4-phenylthiazole hydrobromide monohydrate, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.