missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11483357
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2?-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2′-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10523-68-9
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074743
InChI Key WLDWDRZITJEWRJ-UHFFFAOYSA-N
Synonym 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride
PubChem CID 25331
SMILES [H+].[Cl-].NC1C2CC3CC(C2)CC1C3

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 4297901
Solubility Information Soluble in water.
IUPAC Name adamantan-2-amine;hydrochloride
Formula Weight 187.71
Percent Purity ≥98%
Chemical Name or Material 2-Adamantanamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'