missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylmorpholine, cis + trans, 97%, Thermo Scientific Chemicals

Product Code. 11435730
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-91-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00005973
InChI Key HNVIQLPOGUDBSU-UHFFFAOYSA-N
Synonym morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod
PubChem CID 110862
IUPAC Name 2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Melting Point -85°C
Density 0.935
Boiling Point 147°C
Flash Point 48°C (120°F)
Refractive Index 1.446
Quantity 25 g
UN Number UN1992
Beilstein 103036
Formula Weight 115.18
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylmorpholine, cis + trans
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'