missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxy-p-benzoquinone, 98%, Thermo Scientific Chemicals

Product Code. 11442767
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-55-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00016368
InChI Key OLBNOBQOQZRLMP-UHFFFAOYSA-N
Synonym 2,6-dimethoxy-1,4-benzoquinone, 2,6-dimethoxy-p-benzoquinone, 2,6-dimethoxyquinone, 2,6-dimethoxybenzoquinone, 2,6-dimethoxy-p-quinone, 2,6-dimethoxysemiquinone anions, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethoxy, dmbq, p-benzoquinone, 2,6-dimethoxy, 2,6-dimethoxybenzo-1,4-quinone
PubChem CID 68262
ChEBI CHEBI:27651
IUPAC Name 2,6-dimethoxycyclohexa-2,5-diene-1,4-dione
SMILES COC1=CC(=O)C=C(C1=O)OC

Specifications

Melting Point 252°C to 257°C
Quantity 5 g
Beilstein 2048182
Merck Index 14,3225
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 2,6-Dimethoxy-p-benzoquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'