missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenolindophenol Sodium Salt Hydrate, Honeywell Fluka™

Product Code. 15603910
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Redox indicator

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
PubChem CID 23697355
ChEBI CHEBI:948
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3641229
IUPAC Name sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Formula Weight 290.08g/mol (anhydrous basis)
Loss on Drying ≤14% (120°C)
Odor Odourless
Packaging Glass Bottle
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material 2,6-Dichlorophenolindophenol Sodium Salt Hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'