missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotage™ 2-5mL Reaction Vials
Biotage™ high precision reaction vials are engineered for durability and safe reactions. All vials withstand 30 bar pressures in a wide range of conditions.
Brand: Biotage™ 355630
Code : Z1
Additional Details : Weight : 0.50000kg
Description
- Designed for use with Biotage Initiator+ and Initiator microwave synthesizers
- Designed to support microwave synthesis from mg to g scale without the need for reoptimization
- Designed for safe and efficient heating within its specified volume range
- With the Reseal™ design, the septum can be resealed after it has been penetrated, allowing repeated additions of reagents or in-situ sampling
- Manufactured from contaminant-free microwave-safe glass
- Jump pack of 500 vials
Specifications
Glass | |
Aluminum | |
500 |
2 to 5 mL | |
Septa Cap |