missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)-N,N-dimethylethylamine, 97%, Thermo Scientific™

Product Code. 15450618
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
An intermediate for the synthesis of Tamoxifen.

Solubility
Soluble in chloroform, dichloromethane.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2474-07-9
Molecular Formula C10H14BrNO
Molecular Weight (g/mol) 244.13
MDL Number MFCD00274471
InChI Key MOVOYJFCKMYLHQ-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy-n,n-dimethylethanamine, 2-4-bromophenoxy-n,n-dimethylethylamine, 2-4-bromophenoxy ethyl dimethylamine, 4-2-n,n-dimethylethoxy phenylbromide, 4-2-n,n-dimethylaminoethoxy phenylbromide, 2-4-bromo-phenoxy-ethyl-dimethyl-amine, ethanamine, 2-4-bromophenoxy-n,n-dimethyl, n-2-4-bromophenoxy ethyl-n,n-dimethylamine, 2-4-bromo-phenoxy-ethyl-dimethyl, 4-2-dimethylaminoethoxy bromobenzene
PubChem CID 213877
IUPAC Name 2-(4-bromophenoxy)-N,N-dimethylethanamine
SMILES CN(C)CCOC1=CC=C(C=C1)Br

Specifications

Quantity 5 g
UN Number UN2735
Solubility Information Soluble in chloroform,dichloromethane.
Formula Weight 244.13
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenoxy)-N,N-dimethylethylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'