missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7020927
Click to view available options
Quantity:
100 g
500 g
2500 g
88-05-1
C9H13N
135.21
MFCD00007740
KWVPRPSXBZNOHS-UHFFFAOYSA-N
mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
6913
CHEBI:82545
2,4,6-trimethylaniline
CC1=CC(=C(C(=C1)C)N)C
88-05-1
C9H13N
135.21
MFCD00007740
KWVPRPSXBZNOHS-UHFFFAOYSA-N
mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
6913
CHEBI:82545
2,4,6-trimethylaniline
CC1=CC(=C(C(=C1)C)N)C

2,4,6-Trimethylaniline is useful for the preparation of Grubbs' catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylaniline is useful for the preparation of Grubbs′ catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.

Solubility
Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -5°C
Density 0.962
Boiling Point 232°C to 234°C
Flash Point 96°C (204°F)
Refractive Index 1.551
Quantity 100 g
UN Number UN2810
Beilstein 636559
Solubility Information Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'