missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15451346
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879636-95-0
Molecular Formula C12H11NO3S
Molecular Weight (g/mol) 249.28
MDL Number MFCD06857927
InChI Key WFBWIROXKGGXAW-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-3-methoxyphenyl thiazole-5-carboxylic acid, 2-3-methoxyphenyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-3-methoxy _phenyl-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-3-methoxyphenyl-4-methyl
PubChem CID 4980373
IUPAC Name 2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES COC1=CC=CC(=C1)C1=NC(C)=C(S1)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 249.28
Percent Purity 97%
Chemical Name or Material 2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.