Learn More
2,3-Dichlorothiophene, 97%, Thermo Scientific™
£34.17 - £136.49
Chemical Identifiers
CAS | 17249-79-5 |
---|---|
Molecular Formula | C4H2Cl2S |
Molecular Weight (g/mol) | 153.02 |
InChI Key | LVNVLQIXMBTMPH-UHFFFAOYSA-N |
Synonym | thiophene, 2,3-dichloro, thiophene, dichloro, pubchem15351, acmc-20a4oq, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, 2,3-bis chloranyl thiophene, ksc332o3r |
PubChem CID | 140207 |
IUPAC Name | 2,3-dichlorothiophene |
SMILES | C1=CSC(=C1Cl)Cl |
Description
Chemical Identifiers
17249-79-5 | |
153.02 | |
thiophene, 2,3-dichloro, thiophene, dichloro, pubchem15351, acmc-20a4oq, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, 2,3-bis chloranyl thiophene, ksc332o3r | |
2,3-dichlorothiophene |
C4H2Cl2S | |
LVNVLQIXMBTMPH-UHFFFAOYSA-N | |
140207 | |
C1=CSC(=C1Cl)Cl |
Specifications
17249-79-5 | |
100.0 | |
Colorless to Yellow | |
172°C to 174°C | |
Authentic | |
Glass Bottle | |
1.5687 | |
thiophene, 2,3-dichloro, thiophene, dichloro, pubchem15351, acmc-20a4oq, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, 2,3-bis chloranyl thiophene, ksc332o3r | |
LVNVLQIXMBTMPH-UHFFFAOYSA-N | |
2,3-dichlorothiophene | |
140207 | |
97% | |
2, 3-Dichlorothiophene |
96.0 | |
-37°C | |
1.4700g/mL | |
78°C | |
96% min. (GC) | |
C4H2Cl2S | |
1.47 | |
Solubility in water: <0.2g/L (20 c) | |
C1=CSC(=C1Cl)Cl | |
153.02 | |
153.02 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 2,3-Dichlorothiophene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only