Learn More
2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals
£32.30 - £434.00
Chemical Identifiers
CAS | 513-31-5 |
---|---|
Molecular Formula | C3H4Br2 |
Molecular Weight (g/mol) | 199.87 |
MDL Number | MFCD00000211 |
InChI Key | YMFWYDYJHRGGPF-UHFFFAOYSA-N |
Synonym | 2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712 |
PubChem CID | 10552 |
IUPAC Name | 2,3-dibromoprop-1-ene |
SMILES | BrCC(Br)=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10645962
|
Thermo Scientific Acros
147740100 |
10 mL | Glass bottle |
£32.30
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10447540
|
Thermo Scientific Acros
147740500 |
50 mL | Glass bottle |
£101.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10616944
|
Thermo Scientific Acros
147742500 |
250 mL | Glass bottle |
£434.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-31-5 | |
199.87 | |
YMFWYDYJHRGGPF-UHFFFAOYSA-N | |
10552 | |
BrCC(Br)=C |
C3H4Br2 | |
MFCD00000211 | |
2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712 | |
2,3-dibromoprop-1-ene |
Specifications
513-31-5 | |
100.0 | |
2.1300g/mL | |
81°C | |
75% min. (GC) | |
BrCH2C(Br)=CH2 | |
MFCD00000211 | |
15, 3030 | |
2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712 | |
YMFWYDYJHRGGPF-UHFFFAOYSA-N | |
2,3-dibromoprop-1-ene | |
10552 | |
80% | |
Liquid |
75.0 | |
Yellow to Brown | |
42°C to 44°C (17.0 mmHg) | |
Authentic | |
C3H4Br2 | |
1.5370 to 1.547 | |
01, 201 | |
2.13 | |
Solubility in water: insoluble | |
BrCC(Br)=C | |
199.87 | |
199.87 | |
Technical | |
2, 3-Dibromopropene |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-155-8
- 2,3-Dibromopropene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only