missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™
54.09 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
100 g
Description
C19H15ClN4, CAS Number-298-96-4, 2,3,5-triphenyltetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride, red tetrazolium, tetrazolium red, tetrazolium chloride, uroscreen, urocheck, vitastain, 25g, CHEBI:78019, White, 334.81g/mol
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 235°C |
Color | White |
Packaging | Plastic powder jar |
Quantity | 10 g |
Formula Weight | 334.81g/mol |
Physical Form | Solid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 2,3,5-Triphenyl-2H-tetrazolium chloride
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'