missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Furoin, 98%, Thermo Scientific Chemicals

Product Code. 11482328
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-86-3
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003246
InChI Key MIJRFWVFNKQQDK-UHFFFAOYNA-N
Synonym furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone
PubChem CID 11100
IUPAC Name 1,2-bis(furan-2-yl)-2-hydroxyethanone
SMILES OC(C(=O)C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 137°C to 139°C
Quantity 25 g
Beilstein 383995
Formula Weight 192.17
Percent Purity 98%
Chemical Name or Material 2,2'-Furoin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'