missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dipyridyl disulfide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2,2'-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.
Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.
Notes
Incompatible with oxidizing agents and metals.
2,2′-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.
Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.
Notes
Incompatible with oxidizing agents and metals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 56°C to 59°C |
Flash Point | 210°C (410°F) |
Assay Percent Range | 98% |
Quantity | 5 g |
Beilstein | 154629 |
Solubility Information | Soluble in water,benzene,tetrahydrofuran,dichloromethane,acetonitrile and dimethyformamide. |
Formula Weight | 220.32 |
Percent Purity | 98% |
Chemical Name or Material | 2, 2′-Dipyridyl disulfide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'