Learn More
2,2,6-Trimethylcyclohexanone, 97%, Thermo Scientific Chemicals
£35.60 - £328.00
Chemical Identifiers
CAS | 2408-37-9 |
---|---|
Molecular Formula | C9H16O |
Molecular Weight (g/mol) | 140.226 |
MDL Number | MFCD00045550 |
InChI Key | ZPVOLGVTNLDBFI-UHFFFAOYSA-N |
Synonym | 2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s |
PubChem CID | 17000 |
IUPAC Name | 2,2,6-trimethylcyclohexan-1-one |
SMILES | CC1CCCC(C1=O)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454577
|
Thermo Scientific Alfa Aesar
B21111.03 |
1 g |
£35.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464577
|
Thermo Scientific Alfa Aesar
B21111.06 |
5 g |
£111.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474577
|
Thermo Scientific Alfa Aesar
B21111.14 |
25 g |
£328.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Chemical Identifiers
2408-37-9 | |
140.226 | |
ZPVOLGVTNLDBFI-UHFFFAOYSA-N | |
17000 | |
CC1CCCC(C1=O)(C)C |
C9H16O | |
MFCD00045550 | |
2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s | |
2,2,6-trimethylcyclohexan-1-one |
Specifications
2408-37-9 | |
178°C to 179°C | |
C9H16O | |
MFCD00045550 | |
1905593 | |
Slightly miscible with water. | |
CC1CCCC(C1=O)(C)C | |
140.226 | |
140.23 | |
2,2,6-Trimethylcyclohexanone |
0.904 | |
51°C (125°F) | |
1.447 | |
UN1224 | |
2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s | |
ZPVOLGVTNLDBFI-UHFFFAOYSA-N | |
2,2,6-trimethylcyclohexan-1-one | |
17000 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 219-309-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only