Learn More
1H,1H,2H,2H-Perfluorooctanol, 97%, Thermo Scientific Chemicals
£37.40 - £512.00
Chemical Identifiers
CAS | 647-42-7 |
---|---|
Molecular Formula | C8H5F13O |
Molecular Weight (g/mol) | 364.106 |
MDL Number | MFCD00042143 |
InChI Key | GRJRKPMIRMSBNK-UHFFFAOYSA-N |
Synonym | 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol |
PubChem CID | 69537 |
IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol |
SMILES | C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480701
|
Thermo Scientific Alfa Aesar
B20156.09 |
10 g |
£37.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490701
|
Thermo Scientific Alfa Aesar
B20156.18 |
50 g |
£137.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400711
|
Thermo Scientific Alfa Aesar
B20156.30 |
250 g |
£512.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H,1H,2H,2H-Perfluorooctanol is used to prepare fluorinated aromatic diamine by reacting with 2-(4-nitrophenoxy)-4-nitrophenol through Mitsunobu reaction, which one reacts with dianhydrides such as 4,4′-(hexafluoroisopropylidene)diphthalic anhydride (6FDA), pyromellitic dianhydride (PMDA) and benzophenonetetracarboxylic dianhydride (BTDA) to prepare polyimides.
Solubility
Miscible with chloroform and methanol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
647-42-7 | |
364.106 | |
GRJRKPMIRMSBNK-UHFFFAOYSA-N | |
69537 | |
C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
C8H5F13O | |
MFCD00042143 | |
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol | |
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol |
Specifications
647-42-7 | |
90°C (20 mmHg) | |
C8H5F13O | |
MFCD00042143 | |
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol | |
GRJRKPMIRMSBNK-UHFFFAOYSA-N | |
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol | |
69537 | |
97% |
1.68 | |
91°C (195°F) | |
1.314 | |
1992757 | |
Miscible with chloroform and methanol. Immiscible with water. | |
C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
364.106 | |
364.11 | |
1H,1H,2H,2H-Perfluorooctanol |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 211-477-1
missing translation for 'rtecsNumber' : RH1460000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only