missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Slip-retardant Boot Covers

Catalog Number 15595849
Click to view available options
Size:
Medium
Large
Length (Metric):
30.5 cm
37.5 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Composed of flash-spun HDPE. DuPont™ Tyvek™ IsoClean™ Slip-retardant Boot Covers are permeable to both air and water vapor, yet repel water-based liquids and aerosols. They offer an excellent barrier against fine particles and fibers down to 1μ in size.

  • Fixation ties and slip-retardant sole
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
  • Single packed
  • Note: this product is not CE certified as chemical protective clothing
  • Sterile model available
TRUSTED_SUSTAINABILITY
Size Medium
Material Tyvek Flashspun HDPE
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top with Ankle Ties
Sole Type Anti-skid
Seam Type Bound
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing and Electronics Industries, Other Critical or Controlled Environments
Certifications/Compliance (Europe) Suitable for use in GMP class C/D cleanrooms
Packaging Quantity 100/Cs.
Packaging Type Bulk Packed
Length (Metric) 30.5 cm
Height (Metric) 47cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less

The user should make the risk assessment to determine the protection required.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'