Learn More
Agilent BioTek 284/10 nm Fluorescence Filter
For use with microplate readers and imagers
Brand:  Agilent BioTek 7082259
Additional Details : Weight : 1.00000kg
Product Code. 15321480
  • £680.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
Specifications
Specifications
Microplate Readers and Imagers
284 ±10 nm