missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-1-Phenylethanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Description
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Specifications
Specifications
Melting Point | 19°C to 20°C |
Density | 1.012 |
Boiling Point | 203°C to 205°C |
Quantity | 2500 g |
UN Number | UN2937 |
Solubility Information | Insoluble in water. |
Formula Weight | 122.17 |
Percent Purity | 97% |
Chemical Name or Material | (+/-)-1-Phenylethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'