missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, 99+%, Thermo Scientific Chemicals

Catalog Number p-170910
24.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
90-15-3
C10H8O
144.17
MFCD00003930
KJCVRFUGPWSIIH-UHFFFAOYSA-N
1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
7005
CHEBI:10319
naphthalen-1-ol
C1=CC=C2C(=C1)C=CC=C2O
90-15-3
C10H8O
144.17
MFCD00003930
KJCVRFUGPWSIIH-UHFFFAOYSA-N
1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
7005
CHEBI:10319
naphthalen-1-ol
C1=CC=C2C(=C1)C=CC=C2O

1-Naphthol, 99+%, C10H8O, CAS Number-90-15-3, 1-hydroxynaphthalene, 1-naphthalenol, 1-naphthol, alpha-naphthol, basf ursol ern, fouramine ern, fourrine ern, furro er, naphthol, tertral ern, 100g, 06, 596, 278.0 deg.C to 280.0 deg.C, 100.0, 99.0, CHEBI:10319, Beige-Gray to Brown, 201-969-4, 125 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 95.0°C to 97.0°C
Color Beige-Gray to Brown
Boiling Point 278.0°C to 280.0°C
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C10H7OH
Quantity 100 g
Beilstein 06, 596
Merck Index 15, 6468
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in benzene,alcohol and ether,freely soluble in chloroform,alkali hydroxide,solutions
Formula Weight 144.17
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material 1-Naphthol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Naphthol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'