missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine-4-methanol, 97%, Thermo Scientific™

Product Code. 15404967
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20691-89-8
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD03411903
InChI Key KJZLJGZZDNGGCA-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
PubChem CID 271971
IUPAC Name (1-methylpiperidin-4-yl)methanol
SMILES CN1CCC(CC1)CO

Specifications

Refractive Index 1.476
Quantity 5 g
Formula Weight 129.2
Percent Purity 97%
Chemical Name or Material 1-Methylpiperidine-4-methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'