missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclohexanol, 96%, Thermo Scientific Chemicals

Product Code. 10687472
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-67-0
MDL Number MFCD00003857
InChI Key VTBOTOBFGSVRMA-UHFFFAOYSA-N
Synonym 1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol
PubChem CID 11550
IUPAC Name 1-methylcyclohexan-1-ol
SMILES CC1(CCCCC1)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 26°C
Color Colorless to Yellow
Density 0.9190g/mL
Boiling Point 168°C (752.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C7H14O
Linear Formula CH3C6H10OH
Refractive Index 1.4590 to 1.461
Quantity 5 g
Beilstein 06, 11
Specific Gravity 0.919
Molecular Weight (g/mol) 114.19
Formula Weight 114.19
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 1-Methylcyclohexanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methylcyclohexanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'