missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15460416
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761446-44-0
Molecular Formula C10H17BN2O2
Molecular Weight (g/mol) 208.07
MDL Number MFCD03789259
InChI Key UCNGGGYMLHAMJG-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-4-pyrazoleboronic acid pinacol ester
PubChem CID 2773987
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 59°C to 64°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 208.07
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'