missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible materials are Strong oxidizing agents and Strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible materials are Strong oxidizing agents and Strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 35°C to 36°C (15mmHg) |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 1362893 |
Flash Point | 30°C (86°F) |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 174.32 |
Percent Purity | 97% |
Refractive Index | 1.415 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'