missing translation for 'onlineSavingsMsg'
Learn More

1,4-Anthraquinone, 94%, Thermo Scientific Chemicals

Product Code. 11330114
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-12-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.216
MDL Number MFCD00013724
InChI Key LSOTZYUVGZKSHR-UHFFFAOYSA-N
Synonym 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione
PubChem CID 69457
IUPAC Name anthracene-1,4-dione
SMILES C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1

Specifications

Melting Point ∽218°C (decomposition)
Quantity 5 g
Beilstein 1871197
Solubility Information Insoluble in water.
Formula Weight 208.22
Percent Purity 94%
Chemical Name or Material 1,4-Anthraquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'