missing translation for 'onlineSavingsMsg'
Learn More

1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10456082
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-02-7
Molecular Formula C12H10ClNOS
Molecular Weight (g/mol) 251.73
MDL Number MFCD00068162
InChI Key DLQVOIFYJQATMI-UHFFFAOYSA-N
Synonym 1-3-amino-5-4-chlorophenyl-2-thienyl ethan-1-one, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethanone, 2-acetyl-3-amino-5-4-chlorophenyl thiophene, ethanone,1-3-amino-5-4-chlorophenyl-2-thienyl, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethan-1-one
PubChem CID 2777494
SMILES CC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 1-[3-amino-5-(4-chlorophenyl)thiophen-2-yl]ethanone
Formula Weight 251.74
Percent Purity 97%
Chemical Name or Material 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'