Learn More
1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane, 97%, Thermo Scientific Chemicals
£35.38 - £327.63
Chemical Identifiers
CAS | 2374-14-3 |
---|---|
Molecular Formula | C12H21F9O3Si3 |
Molecular Weight (g/mol) | 468.538 |
MDL Number | MFCD00054667 |
InChI Key | URZHQOCYXDNFGN-UHFFFAOYSA-N |
Synonym | silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer |
PubChem CID | 75410 |
IUPAC Name | 2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane |
SMILES | C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319325
|
Thermo Scientific Alfa Aesar
L16680.06 |
5 g |
£35.38
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329325
|
Thermo Scientific Alfa Aesar
L16680.14 |
25 g |
£116.58
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339325
|
Thermo Scientific Alfa Aesar
L16680.22 |
100 g |
£327.63
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2374-14-3 | |
468.538 | |
URZHQOCYXDNFGN-UHFFFAOYSA-N | |
75410 | |
C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F |
C12H21F9O3Si3 | |
MFCD00054667 | |
silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer | |
2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane |
Specifications
135°C (22mmHg) | |
2374-14-3 | |
MFCD00054667 | |
1809096 | |
URZHQOCYXDNFGN-UHFFFAOYSA-N | |
2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane | |
75410 | |
97% | |
1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane |
34°C to 36°C | |
C12H21F9O3Si3 | |
UN2811 | |
silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer | |
C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F | |
468.538 | |
468.54 | |
1.24 |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P281-P308+P313-P501c
H361-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 219-154-7
missing translation for 'rtecsNumber' : GZ4818000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only