missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Trifluoromethoxy)phenyl]ethanol, 97%, Thermo Scientific™

£49.00 - £140.00

Chemical Identifiers

CAS 910442-37-4
Molecular Formula C9H9F3O2
Molecular Weight (g/mol) 206.164
MDL Number MFCD08062367
InChI Key LWOXLMRGAQEYMM-UHFFFAOYSA-N
Synonym 1-2-trifluoromethoxy phenyl ethanol, 1-2-trifluoromethoxy phenyl ethan-1-ol, 1-2-trifluoromethoxyphenyl ethanol
PubChem CID 44891073
IUPAC Name 1-[2-(trifluoromethoxy)phenyl]ethanol
SMILES CC(C1=CC=CC=C1OC(F)(F)F)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484937
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32244.06
5 g
£49.00
5g
Please to purchase this item. Need a web account? Register with us today!
15494937
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32244.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

910442-37-4
206.164
LWOXLMRGAQEYMM-UHFFFAOYSA-N
44891073
CC(C1=CC=CC=C1OC(F)(F)F)O
C9H9F3O2
MFCD08062367
1-2-trifluoromethoxy phenyl ethanol, 1-2-trifluoromethoxy phenyl ethan-1-ol, 1-2-trifluoromethoxyphenyl ethanol
1-[2-(trifluoromethoxy)phenyl]ethanol

Specifications

910442-37-4
MFCD08062367
Insoluble in water.
CC(C1=CC=CC=C1OC(F)(F)F)O
206.164
206.16
1-[2-(Trifluoromethoxy)phenyl]ethanol
C9H9F3O2
1-2-trifluoromethoxy phenyl ethanol, 1-2-trifluoromethoxy phenyl ethan-1-ol, 1-2-trifluoromethoxyphenyl ethanol
LWOXLMRGAQEYMM-UHFFFAOYSA-N
1-[2-(trifluoromethoxy)phenyl]ethanol
44891073
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications