missing translation for 'onlineSavingsMsg'
Learn More
1-(2-Fluoro-4-nitrophenyl)azepane, 97%, Thermo Scientific™ Product Code.: 15429938

1-(2-Fluoro-4-nitrophenyl)azepane, 97%, Thermo Scientific™

Product Code. 15429938
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15429938

Brand: Thermo Scientific Alfa Aesar H63305.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 250371-80-3
Molecular Formula C12H15FN2O2
Molecular Weight (g/mol) 238.262
InChI Key LJVYPBVHNCEVFI-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-nitrophenyl azepane
PubChem CID 2872204
IUPAC Name 1-(2-fluoro-4-nitrophenyl)azepane
SMILES C1CCCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

Quantity 1 g
UN Number UN3259
Formula Weight 238.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)azepane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(2-Fluoro-4-nitrophenyl)azepane, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.