Learn More
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
£18.60 - £722.00
Chemical Identifiers
CAS | 106-93-4 |
---|---|
Molecular Formula | C2H4Br2 |
Molecular Weight (g/mol) | 187.86 |
InChI Key | PAAZPARNPHGIKF-UHFFFAOYSA-N |
Synonym | ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom |
PubChem CID | 7839 |
ChEBI | CHEBI:28534 |
IUPAC Name | 1,2-dibromoethane |
SMILES | C(CBr)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10695142
|
Thermo Scientific Acros
112790250 |
25 mL | Glass bottle |
£18.60
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10244520
|
Thermo Scientific Acros
112795000 |
500 mL | Glass bottle |
£67.20
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10346270
|
Thermo Scientific
112790010 |
1 L | Glass bottle |
£87.85
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974481
|
Thermo Scientific Acros
112790025 |
2.5 L | Plastic Bottle |
£192.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15199153
|
Thermo Scientific Acros
112790100 |
10 L | Plastic Drum |
£722.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-93-4 | |
187.86 | |
ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom | |
CHEBI:28534 | |
C(CBr)Br |
C2H4Br2 | |
PAAZPARNPHGIKF-UHFFFAOYSA-N | |
7839 | |
1,2-dibromoethane |
Specifications
106-93-4 | |
100.0 | |
Colorless to Yellow | |
131.0°C to 132.0°C | |
Authentic | |
C2H4Br2 | |
1.5375 to 1.5395 | |
01,374; 04,126; 05,187; 12,570 | |
2.173 | |
Solubility in water: 4g/L (20°C). Other solubilities: miscible with most organic solvents | |
C(CBr)Br | |
187.86 | |
CHEBI:28534 | |
99% | |
1,2-Dibromoethane |
98.5 | |
9.0°C to 10.0°C | |
2.1730g/mL | |
>104°C | |
98.5% min. (GC) | |
BrCH2CH2Br | |
01,9 | |
15,385 | |
ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom | |
PAAZPARNPHGIKF-UHFFFAOYSA-N | |
1,2-dibromoethane | |
7839 | |
187.86 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause cancer.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-444-5
RUO – Research Use Only