missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Cyclohexanedione, 98%, Thermo Scientific Chemicals
£110.00 - £1418.00
Chemical Identifiers
CAS | 765-87-7 |
---|---|
Molecular Formula | C6H8O2 |
Molecular Weight (g/mol) | 112.13 |
InChI Key | OILAIQUEIWYQPH-UHFFFAOYSA-N |
Synonym | 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol |
PubChem CID | 13006 |
ChEBI | CHEBI:41674 |
IUPAC Name | cyclohexane-1,2-dione |
SMILES | C1CCC(=O)C(=O)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10636872
|
Thermo Scientific Acros
111160100 |
10 g | Glass bottle |
£110.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10385980
|
Thermo Scientific
111160500 |
50 g | Glass bottle |
£286.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974851
|
Thermo Scientific Acros
111162500 |
250 g | Glass Bottle |
£1418.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
765-87-7 | |
112.13 | |
1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
CHEBI:41674 | |
C1CCC(=O)C(=O)C1 |
C6H8O2 | |
OILAIQUEIWYQPH-UHFFFAOYSA-N | |
13006 | |
cyclohexane-1,2-dione |
Specifications
765-87-7 | |
Yellow | |
84°C | |
97.5% min. (GC) | |
C6H8(=O)2 | |
07,II,526 | |
1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
OILAIQUEIWYQPH-UHFFFAOYSA-N | |
cyclohexane-1,2-dione | |
13006 | |
112.13 | |
Liquid After Melting |
35.0°C to 38.0°C | |
193.0°C to 195.0°C | |
Authentic | |
C6H8O2 | |
1.5180 to 1.5200 | |
02,93 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol and diethyl ether | |
C1CCC(=O)C(=O)C1 | |
112.13 | |
CHEBI:41674 | |
98% | |
1, 2-Cyclohexanedione |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-155-3
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,2-Cyclohexanedione
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only