missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF169 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26344-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF169 (Zinc finger protein 169) is a 603 amino acid nuclear protein that contains one KRAB domain and thirteen C2H2-type zinc fingers. ZNF169 is highly expressed in kidney and weakly expressed in spleen, liver, small intestine and heart, where it functions as a transcription regulator. The gene encoding ZNF169 maps to a region of human chromosome 9q22, which has been associated with many human diseases such as colon cancer, migraine auras, basal cell carcinoma, Gorlin syndrome and Extraskeletal myxoid chondrosarcoma.Specifications
ZNF169 | |
Polyclonal | |
Unconjugated | |
ZNF169 | |
zinc finger protein 169, ZNF169 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
169841, 67911 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14929 | |
Zfp169, ZNF169 | |
ZNF169 Fusion Protein Ag23805 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |