missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, Reagent grade, ≥98%, Honeywell™
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 293°C |
Color | White |
Physical Form | Solid |
Quantity | 5 g |
Linear Formula | ZnCl2 |
UN Number | UN2331 |
Formula Weight | 136.3g/mol |
Percent Purity | ≥98% |
Grade | Reagent |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'