missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Reagent grade, ≥98%, Honeywell™

Product Code. 15641820
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Melting Point 293°C
Color White
Physical Form Solid
Quantity 500 g
Linear Formula ZnCl2
UN Number UN2331
Formula Weight 136.3g/mol
Percent Purity ≥98%
Grade Reagent
Packaging Poly Bottle
Vapor Pressure 1mmHg (428°C)
Density 2.93g/cu.cm
Chemical Name or Material Zinc Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'