missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR86 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548389
Additional Details : Weight : 0.01000kg
Description
WDR86 Polyclonal antibody specifically detects WDR86 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
WDR86 | |
Polyclonal | |
Unconjugated | |
WDR86 | |
WD repeat domain 86; WD repeat-containing protein 86; WDR86 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
349136 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q86TI4 | |
WDR86 | |
KLH conjugated synthetic peptide between 286-316 amino acids from the C-terminal region of human WDR86. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |