missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR83 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560143
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CLSERDTHVV SCSEDGKVFF WDLVEGALAL ALPVGSGVVQ SLAYHPTEPC LLTAMGGSVQ CWREEAYEAE DGAG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
WDR83 Polyclonal antibody specifically detects WDR83 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
WDR83 | |
Polyclonal | |
Unconjugated | |
WDR83 | |
1500041N16Rik; MAPK cascade scaffold protein; MAPK organizer 1; MGC4238; Mitogen-activated protein kinase organizer 1; MORG1; RGD1306947; WD repeat domain 83; WD repeat domain containing 83; WD repeat domain-containing protein 83; WD repeat protein Morg1; WDR83 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRX9 | |
WDR83 | |
Recombinant protein corresponding to Human WDR83 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only