Learn More
WDR13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551483
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GHRRSVSRGS YQLQAQMNRA VYEDRPPGSV VPTSAAEASR AMAGDTSLSE NYAFAGMYHV FDQHVDEAVP RVRFANDDRH RLACCSLDGS ISLCQLVPAP PTVLRVLR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
WDR13 Polyclonal antibody specifically detects WDR13 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
WDR13 | |
Polyclonal | |
Unconjugated | |
WDR13 | |
1700060B08Rik; 5730411P10Rik; DXHXS7467e; LOW QUALITY PROTEIN: WD repeat-containing protein 13; memory-related protein; MG21; mMg21; RGD1560982; W51679; WD repeat domain 13; WD repeat domain 13 protein; WD repeat-containing protein 13; Wdr13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64743 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1Z4 | |
WDR13 | |
Recombinant protein corresponding to Human WDR13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only