missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDFY4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA568444
Additional Details : Weight : 0.01000kg
Description
WDFY4 Polyclonal antibody specifically detects WDFY4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
WDFY4 | |
Polyclonal | |
Unconjugated | |
Wdfy4 | |
C10orf64; Gm18810; KIAA1607; RGD1564142; WD repeat and FYVE domain containing 4; WD repeat- and FYVE domain-containing protein 4; WDFY family member 4; Wdfy4 | |
Rabbit | |
affinity chromatography | |
RUO | |
57705 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6ZS81 | |
Wdfy4 | |
Fusion protein of human WDFY4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |