Learn More
Watson-Marlow 313 Rapid-Load Pumphead Extension
Offers simple, error-free tube loading
Brand: Watson-Marlow 033.3431.000
Code : 50
Additional Details : Weight : 0.11000kg
Description
- Pumphead accepts seven tube sizes to meet any flow requirement
- Three nylatron rollers
- Tube wall of 1.6mm
- With adjustable tube clamps
Specifications
Metering and transfer pump | |
Adjustable tube clamps | |
Extension Pump Head |
3 | |
Nylatron |