missing translation for 'onlineSavingsMsg'
Learn More

Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals

Catalog Number 11368078
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Used in conductometric titration, Synthesis of nanomaterials. Mostly it is used as solvent in industries and in all research fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Solubility Information Miscible in methanol,acetone,Ethanol.
Formula Weight 18.02
Grade ACS
Odor Odorless
Density 1
Quantity 1 L
Refractive Index 1.333
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'