Learn More
VIPAS39 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5110569
Additional Details : Weight : 0.01000kg
Description
Sequence of this protein is as follows: MNRTKGDEEE YWNSSKFKAF TFDDEDDELS QLKESKRAVN SLRDFVDDDD DDDLERVSWS GEPVGSISWS IRETAGNSGS THEGREQLKS RNSFSSYAQL PKPTSTYSLS SFFRGRTRPG SFQSLSDALS DTPAKSYAPE LGRPKGEYRD YSNDWSPSDT VRRLRKGKVC SLERFRSLQD KLQLLEEAVS MHDGNVITAV LIFLKRTLSK EILFRELEVR
VIPAS39 Polyclonal antibody specifically detects VIPAS39 in Human, Mouse samples. It is validated for Western Blot.Specifications
VIPAS39 | |
Polyclonal | |
Unconjugated | |
VIPAS39 | |
6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction | |
Rabbit | |
Affinity Chromatography | |
RUO | |
104799, 63894 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.05 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGQ1, Q9H9C1 | |
VIPAS39 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human VIPAS39 (NP_071350.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |