missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | VAMP8 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894764
|
Proteintech
15546-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884764
|
Proteintech
15546-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.Specifications
VAMP8 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O70404, Q9BV40, Q9WUF4 | |
22320, 83730, 8673 | |
VAMP8 Fusion Protein Ag7903 | |
Primary | |
-20°C | |
VAMP8 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EDB, Endobrevin, VAMP 8, VAMP8 | |
VAMP8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |