Learn More
Edwards Vacuum™ XDS5/10 Tip Seal and Exhaust Kit
For use with Dry Scroll Vacuum Pumps.
Brand:  Edwards Vacuum™ A72601805
Code : 03
Additional Details : Weight : 1.00000kg
Product Code. 11209839
  • £496.02 / Each
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
Description
Description
  • Replacement set
  • OEM part
  • Includes O-rings, tip seals and exhaust valve for the Edwards Vacuum™ XDS 5 and XCS 10 Dry Scroll Vacuum Pumps
Order Info
Type
Specifications
Specifications
XDS5,XDS5C,XDS10,XDS10C
Includes O-rings, tip seals and exhaust valve
1 Each
Vacuum Pump Tip Seal Replacement Kit