Learn More
BUCHI V-300 Vacuum Pump
Use this pump in with the I-300 or I-300 Pro Interface for economical, speed-controlled operation and low noise emission. BUCHI V-300 Vacuum Pumps feature chemically-resistant PTFE construction and a transparent front panel. Optimally suited to work with the Rotavapor R-300.
Brand:  BUCHI 11V300000
2936.30 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Product Code. 15511280
  • £3090.84 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chemically resistant PTFE diaphragm pump
  • Seamless plug & play integration with the I-300/I-300 Pro Interface and the fully automated Rotavapor R-300 System
  • Contaminants are easily visible through transparent front panel
  • Unattended operation in combination with Interface I-300/I-300 Pro
  • Optional level sensor for secondary condenser alerts potential overflow of the receiving flask
  • High chemical resistance due to the use of high quality materials
  • Quiet operation and increased lifetime thanks to speed-controlled operation in conjunction with the optional Interface I-300/I-300 Pro
Specifications
Specifications
5 mbar (±2 mbar)
1500 rpm max.
100/240 V 50/60 Hz
321 mm
11V300000
60 to 180 w
1.8 m³/h
291 mm
General laboratory vacuum generation
Vacuum tubing, power cable, silencer, tubing connection set, mini-DIN communication cable, Membrane tool (to fasten and remove membrane)
CE / CSA / CUS
BUCHI V-300 Vacuum Pump, Stand-alone
PTFE
32 to 57 dBA (depending on operating mode); 32 dBA: with 10% load (normal); 57 dBA: with 100% load
7.6 kg
50/60 Hz
200 mm
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products