Learn More
USP34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555261
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QHIIPFRTYV IRYLCKLSDQ ELRQSAARNM ADLMWSTVKE PLDTTLCFDK ESLDLAFKYF MSPTLTMRLA GLSQITNQLH TFNDVCNNES LVSDTETSIA KELADWLI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 21%.
USP34 Polyclonal antibody specifically detects USP34 in Human samples. It is validated for Immunohistochemistry.Specifications
USP34 | |
Polyclonal | |
Unconjugated | |
USP34 | |
A530081C03Rik; deubiquitinating enzyme 34; KIAA0570; KIAA0729; Murr2; U2af1-rs1 region 2; ubiquitin carboxyl-terminal hydrolase 34; ubiquitin specific peptidase 34; ubiquitin specific protease 34; ubiquitin thioesterase 34; ubiquitin thiolesterase 34; ubiquitin-specific-processing protease 34; Usp34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9736 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q70CQ2 | |
USP34 | |
Recombinant protein corresponding to Human USP34 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only