Learn More
URB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553787
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VEHHKTCRSL GRSLWQQPSV GDILRLLDRD RMMQTILHFP QNRRLLPPED TQELIFKDKS RVDLDGLYDP CFLLQLFSEL TRPEFVVDCR KFLDSNALGL TVTALSSYDP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
URB1 Polyclonal antibody specifically detects URB1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
URB1 | |
Polyclonal | |
Unconjugated | |
URB1 | |
4921511H13Rik; 5730405K23Rik; AA670781; AI645590; C21orf108; KIAA0539; mKIAA0539; NOP254; NPA1; nucleolar pre-ribosomal-associated protein 1; nucleolar preribosomal-associated protein 1; nucleolar protein 254 kDa; Protein URB1; URB1; URB1 ribosome biogenesis 1 homolog; URB1 ribosome biogenesis 1 homolog (S. cerevisiae) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60287 | |
URB1 | |
Recombinant protein corresponding to Human URB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only