missing translation for 'onlineSavingsMsg'
Learn More

UQCRC2 Mouse anti-Human, Unconjugated, Clone: 1A5G11, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen UQCRC2
Clone 1A5G11
Concentration 500 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16880236
View Documents
Proteintech
67547-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16870236
View Documents
Proteintech
67547-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The UQCRC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UQCRC2. This antibody recognizes human, mouse, rat, pig antigen. The UQCRC2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

UQCRC2
500 μg/mL
Monoclonal
Mouse
7385
Primary
1A5G11
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Human, Mouse, Rat, Pig
IgG1
Videos
SDS
Documents

Documents

Product Certifications