Learn More
Smeg™ Universal Basket
Used with standard baskets CS1-1 and CS2 . Smeg Instruments Universal Basket is used with trolley LM20.
Brand: Smeg™ SB30
This item is not returnable.
View return policy
Description
- 30-position support to hold beakers of any size
- Has capacity of 30 items
- Made of stainless steel
- Recommended for beakers with capacity smaller than 500mL
Specifications
Universal Basket |