Learn More
UGGT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552981
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DFILSHAVYC RDVLKLKKGQ RAVISNGRII GPLEDSELFN QDDFHLLENI ILKTSGQKIK SHIQQLRVEE DVASDLVMKV DALLSAQPKG DPRIEYQFFE DRHSAIKLRP KEGETYFDVV AVVDPV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
UGGT1 Polyclonal antibody specifically detects UGGT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
UGGT1 | |
Polyclonal | |
Unconjugated | |
Uggt1 | |
0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; Gt; HUGT1; RUGT; rUGT1; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein glucosyltransferase 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGCGL1; Uggt; Uggt1; Ugt1; Ugtr | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171129, 320011, 56886 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P5E4, Q9JLA3, Q9NYU2 | |
Uggt1 | |
Recombinant protein corresponding to Human UGGT1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only