Learn More
UGCG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554986
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLHLNKKATD KQPYSKLPGV SLLKPLKGVD PNLINNLETF FELDYPKYEV LLCVQDHDDP AIDVCKKLLG KYPNVDARLF IGGKKVGINP KINNLMPGYE VAKYDLI Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
UGCG Polyclonal antibody specifically detects UGCG in Human samples. It is validated for Immunohistochemistry.Specifications
UGCG | |
Polyclonal | |
Unconjugated | |
UGCG | |
AU043821; C80537; cb539; Ceramide glucosyltransferase; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 21; Epcs21; GCS; GlcT; GLCT1; GLCT-1; glucosylceramide synthase; hypothetical protein LOC553944; sb:cb539; UDP-glucose ceramide glucosyltransferase; UDP-glucose:ceramide glycosyltransferase; UDP-glucose:N-acylsphingosine D-glucosyltransferase; Ugcg; Ugcgl; zgc:112506 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16739 | |
UGCG | |
Recombinant protein corresponding to Human UGCG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only